Merck
  • A lactose fermentation product produced by Lactococcus lactis subsp. lactis, acetate, inhibits the motility of flagellated pathogenic bacteria.
A lactose fermentation product produced by Lactococcus lactis subsp. lactis, acetate, inhibits the motility of flagellated pathogenic bacteria.
Microbiology (Reading, England) (2015-01-13)
Shuichi Nakamura, Yusuke V Morimoto, Seishi Kudo
ABSTRACT
Many strains of lactic acid bacteria have been used for the production of probiotics. Some metabolites produced by lactic acid bacteria impair the motilities of pathogenic bacteria. Because bacterial motility is strongly associated with virulence, the metabolic activities of lactic acid bacteria are effective for suppressing bacterial infections. Here we show that lactose fermentation by Lactococcus lactis subsp. lactis inhibits the motility of Salmonella enterica serovar Typhimurium. A single-cell tracking and rotation assay for a single flagellum showed that the swimming behaviour of Salmonella was severely but transiently impaired through disruption of flagellar rotation on exposure to media cultivated with Lac. lactis. Using a pH-sensitive fluorescent protein, we observed that the intracellular pH of Salmonella was decreased because of some fermentation products of Lac. lactis. We identified acetate as the lactose fermentation product of Lac. lactis triggering the paralysis of Salmonella flagella. The motilities of Pseudomonas, Vibrio and Leptospira strains were also severely disrupted by lactose utilization by Lac. lactis. These results highlight the potential use of Lac. lactis for preventing infections by multiple bacterial species.
Sigma-Aldrich
D-Lactose monohydrate, Vetec, reagent grade
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)