- Fluorination of thiocarbonyl compounds with Bis(2-methoxyethyl)aminosulfur trifluoride (Deoxo-fluor reagent): A facile synthesis of gem-difluorides
Fluorination of thiocarbonyl compounds with Bis(2-methoxyethyl)aminosulfur trifluoride (Deoxo-fluor reagent): A facile synthesis of gem-difluorides
The Journal of organic chemistry (2000-08-24)
Lal, Lobach, Evans
PMID10956459
ABSTRACT
A variety of thiocarbonyl derivatives (thioketone, thioester, thioamide, dithioester, and dithiocarbamate) were converted to the corresponding gem-difluorides in excellent yields on reaction with the fluorinating agent, bis(2-methoxyethyl)aminosulfur trifluoride, in the presence of SbCl(3).