Merck
  • LGR5 Activates Noncanonical Wnt Signaling and Inhibits Aldosterone Production in the Human Adrenal.
LGR5 Activates Noncanonical Wnt Signaling and Inhibits Aldosterone Production in the Human Adrenal.
The Journal of clinical endocrinology and metabolism (2015-04-29)
Lalarukh Haris Shaikh, Junhua Zhou, Ada E D Teo, Sumedha Garg, Sudeshna Guha Neogi, Nichola Figg, Giles S Yeo, Haixiang Yu, Janet J Maguire, Wanfeng Zhao, Martin R Bennett, Elena A B Azizan, Anthony P Davenport, Grahame McKenzie, Morris J Brown
ABSTRACT
Aldosterone synthesis and cellularity in the human adrenal zona glomerulosa (ZG) is sparse and patchy, presumably due to salt excess. The frequency of somatic mutations causing aldosterone-producing adenomas (APAs) may be a consequence of protection from cell loss by constitutive aldosterone production. The objective of the study was to delineate a process in human ZG, which may regulate both aldosterone production and cell turnover. This study included a comparison of 20 pairs of ZG and zona fasciculata transcriptomes from adrenals adjacent to an APA (n = 13) or a pheochromocytoma (n = 7). Interventions included an overexpression of the top ZG gene (LGR5) or stimulation by its ligand (R-spondin-3). A transcriptome profile of ZG and zona fasciculata and aldosterone production, cell kinetic measurements, and Wnt signaling activity of LGR5 transfected or R-spondin-3-stimulated cells were measured. LGR5 was the top gene up-regulated in ZG (25-fold). The gene for its cognate ligand R-spondin-3, RSPO3, was 5-fold up-regulated. In total, 18 genes associated with the Wnt pathway were greater than 2-fold up-regulated. ZG selectivity of LGR5, and its absence in most APAs, were confirmed by quantitative PCR and immunohistochemistry. Both R-spondin-3 stimulation and LGR5 transfection of human adrenal cells suppressed aldosterone production. There was reduced proliferation and increased apoptosis of transfected cells, and the noncanonical activator protein-1/Jun pathway was stimulated more than the canonical Wnt pathway (3-fold vs 1.3-fold). ZG of adrenal sections stained positive for apoptosis markers. LGR5 is the most selectively expressed gene in human ZG and reduces aldosterone production and cell number. Such conditions may favor cells whose somatic mutation reverses aldosterone inhibition and cell loss.
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Roche
Digoxigenin-11-dUTP, alkali-stable
Sigma-Aldrich
MISSION® esiRNA, targeting human LGR5
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lgr5