Merck
  • A Cascade of epistatic interactions regulating teratozoospermia in mice.
A Cascade of epistatic interactions regulating teratozoospermia in mice.
Mammalian genome : official journal of the International Mammalian Genome Society (2015-05-13)
Keitaro Hirawatari, Naoto Hanzawa, Ikuo Miura, Shigeharu Wakana, Hideo Gotoh
ABSTRACT
Infertility in humans and subfertility in domestic animals are two major reproductive problems. Among human couples, ~15% are diagnosed as infertile, and males are considered responsible in about 50% of the cases. To examine male fertility, various sperm tests including analyses of sperm morphology, sperm count and sperm mobility are usually performed. Teratozoospermia, a condition characterized by the presence of morphologically abnormal sperm, is considered as a symptom of infertility. B10.MOL-TEN1 (TEN1) mice (Mus musculus) show inherited teratozoospermia at high frequencies (~50%). In this study, the polygenic control of teratozoospermia in the TEN1 strain was analysed. A quantitative trait loci analysis indicated three statistically significant loci, Sperm-head morphology 3 (Shm3; logarithm of the odds (LOD) score, 29.25), Shm4 (LOD score, 6.80), and Shm5 (LOD score, 3.58). These three QTL peaks were mapped to 24.3 centimorgans (cM) on chromosome 1, 32.0 cM on chromosome X, and 63.8 cM on chromosome 6, respectively. Another locus that is yet to be determined was also predicted. Shm3 was found to be the major locus responsible for teratozoospermia, and a sequential cascade of interactions of the other three loci was apparent. These results are expected to help understand the mechanisms underlying reproductive problems in humans or domestic animals.
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium acetate, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)