Merck
  • Dimethyl fumarate suppresses Theiler's murine encephalomyelitis virus-induced demyelinating disease by modifying the Nrf2-Keap1 pathway.
Dimethyl fumarate suppresses Theiler's murine encephalomyelitis virus-induced demyelinating disease by modifying the Nrf2-Keap1 pathway.
International immunology (2015-02-28)
Kunitoshi Kobayashi, Hiroki Tomiki, Yuji Inaba, Motoki Ichikawa, Byung S Kim, Chang-Sung Koh
ABSTRACT
Dimethyl fumarate (DMF) is a modifier of the nuclear factor (erythroid-derived 2)-2 (Nrf2)-kelch-like ECH-associated protein 1 (Keap1) pathway. DMF treatment in the effector phase significantly suppressed the development of Theiler's murine encephalomyelitis virus-induced demyelinating disease (TMEV-IDD) both clinically and histologically. DMF treatment leads to an enhanced Nrf2 antioxidant response in TMEV-IDD mice. DMF treatment in the effector phase significantly suppressed the level of IL-17A mRNA. DMF is known to inhibit differentiation of T helper 17 (Th17) cells via suppressing NF-κB. Taken together, our data suggest that DMF treatment in the effector phase may suppress TMEV-IDD not only via enhancing the antioxidant response but also via suppressing IL-17A.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptprc