Merck
  • Effective removal of cationic dyes from aqueous solution using gum ghatti-based biodegradable hydrogel.
Effective removal of cationic dyes from aqueous solution using gum ghatti-based biodegradable hydrogel.
International journal of biological macromolecules (2015-05-03)
Hemant Mittal, Arjun Maity, Suprakas Sinha Ray
ABSTRACT
Biodegradable hydrogels of gum ghatti (Gg) with a co-polymer mixture of acrylamide (AAm) and methacrylic acid (MAA) (termed as Gg-cl-P(AAm-co-MAA)) were synthesised by microwave-assisted free radical graft co-polymerisation technique. The hydrogel polymer was characterized by FTIR, SEM, and Brunauer-Emmett-Teller techniques. The Gg-cl-P(AAm-co-MAA) hydrogel was studied as an adsorbent for the removal of methylene blue (MB) and methyl violet (MV) from aqueous solutions. Adsorption of both the dyes followed pseudo-second-order kinetics and Langmuir adsorption isotherm models. The hydrogel polymer adsorbed 98% of MB and 95% of MV from aqueous solution. The Gg-cl-P(AAm-co-MAA) maintained its original sorption capacity for three cycles of adsorption-desorption. Furthermore, the hydrogel polymer degraded fully within 50 days in soil compost. In summary, the Gg-cl-P(AAm-co-MAA) hydrogel could be a potential adsorbent for the remediation of dyes from industrial wastewater.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methyl acetoacetate, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)