Merck
  • Local administration of stromal cell-derived factor-1 promotes stem cell recruitment and bone regeneration in a rat periodontal bone defect model.
Local administration of stromal cell-derived factor-1 promotes stem cell recruitment and bone regeneration in a rat periodontal bone defect model.
Materials science & engineering. C, Materials for biological applications (2015-06-05)
Hongrui Liu, Minqi Li, Lingqian Du, Pishan Yang, Shaohua Ge
ABSTRACT
Stromal cell-derived factor-1 (SDF-1) recruits adult stem/progenitor cells via its specific receptor, C-X-C motif receptor 4 (CXCR4), to promote heart, kidney and tendon regeneration, but little is known about the effects of SDF-1 on bone regeneration in periodontal diseases. The objective of this study was to investigate whether local administration of SDF-1 in a collagen membrane scaffold enhanced the recruitment of host stem cells and improved periodontal bone defect repair. To this end, bone defects were established on the buccal side of bilateral mandibles in Wistar rats. After application of collagen membranes loaded with SDF-1 or phosphate-buffered saline (PBS) to the defects, the effects of SDF-1 on stem cell recruitment, inflammatory cell responses, angiogenesis, osteoclastogenesis, scaffold degradation, and bone regeneration were evaluated. It showed that SDF-1 recruited host-derived mesenchymal stem cells and hematopoietic stem cells to the wound area and significantly reduced the CD11b+ inflammatory cell response. Moreover, SDF-1 increased vascular formation, induced early bone osteoclastogenesis, accelerated scaffold degradation, and promoted the quality and quantity of regenerated bone. Our results suggest that this cell-free approach by local administration of SDF-1 may be an effective strategy for development as a simple and safe technique for periodontal bone regeneration.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet