Merck
  • Nanoparticle and polysaccharide conjugate: a potential candidate vaccine to improve immunological stimuli.
Nanoparticle and polysaccharide conjugate: a potential candidate vaccine to improve immunological stimuli.
International journal of biological macromolecules (2014-12-03)
K Sanjana P Devi, Banalata Sahoo, Birendra Behera, Tapas K Maiti
ABSTRACT
Active polysaccharides isolated from various fungal sources have been implicated to stimulate immune response against various pathogens as well as self anomalies such as cancer. Therefore, the nuanced approach presented in our work was to blend polysaccharides derived from Pleurotus ostreatus with biocompatible ferrite nanoparticles and thereafter investigate the enhanced immune functionality of the polysaccharide-nanoparticle composite. A Schiff base reductive amination reaction occurred between the aldehyde group of the polysaccharide and the amine group of the nanoparticles in the presence of a strong reducing agent such as sodium cyanoborohydride to form a stable amide bond between the two conjugating molecules. The multifaceted conjugate was characterized by physiochemical techniques such as electron microscopy, FTIR, VSM and DLS measurements. This particulate form of the polysaccharide showed a marked escalation in the production of free radicals such as reactive oxygen and nitrogen species in murine macrophages as compared to the soluble form. Animal based experiments demonstrated a reduction in tumor volume and augmentation in the proliferation of splenocytes in particulate or conjugated polysaccharide treated mice. Furthermore, molecular signaling studies showed a high upregulation in p-p38 and p-MEK molecules in particulate polysaccharide treated RAW264.7 cells suggesting a cellular downstream mechanistic regulation behind the immunostimulative response.
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Hematoxylin