Merck
  • New insight on optical and magnetic Fe3O4 nanoclusters promising for near infrared theranostic applications.
New insight on optical and magnetic Fe3O4 nanoclusters promising for near infrared theranostic applications.
Nanoscale (2015-07-08)
Chih-Chia Huang, Po-Yang Chang, Chien-Liang Liu, Jia-Pu Xu, Shu-Pao Wu, Wen-Chuan Kuo
ABSTRACT
Extensive efforts have been devoted to the development of a new biophotonic system using near infrared (NIR) nano-agents for non-invasive cancer diagnosis and therapy. Here, we developed a simple synthesis reaction of ligands, hydrazine, and iron(ii) chloride to fabricate Fe3O4 cluster-structured nanoparticles (CNPs) with interesting NIR photonics and high magnetization (Ms: 98.3 emu g(-1) and proton relaxivity r2: 234.6 mM(-1) s(-1)). These Fe3O4 CNPs exhibited optical absorption and reflection over all wavelengths, showing a U-shape absorption band with a low absorbance at a range of 750-950 nm and a progressive evolution in the second near infrared region. Strengthening of the scattering effect by incubating Fe3O4 CNPs with HeLa cells was observed when optical contrast enhancement was performed in an optical coherence tomography (OCT) microscope system with a laser light source at 860 nm. Using a 1064 nm laser at a low power density (380 mW cm(-2)) to excite the Fe3O4 CNPs (375 ppm[Fe]) led to a rise in the water temperature from 25 °C to 58 °C within 10 min. Finally, we present the first example of magnetomotive OCT cellular imaging combined with enhanced photothermal therapy using Fe3O4 CNPs and applying a magnetic field, which is promising for preclinical and clinical trials in the future.
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%