Merck
Membranes with artificial free-volume for biofuel production.
Nature communications (2015-06-25)
Nikos Petzetakis, Cara M Doherty, Aaron W Thornton, X Chelsea Chen, Pepa Cotanda, Anita J Hill, Nitash P Balsara
ABSTRACT
Free-volume of polymers governs transport of penetrants through polymeric films. Control over free-volume is thus important for the development of better membranes for a wide variety of applications such as gas separations, pharmaceutical purifications and energy storage. To date, methodologies used to create materials with different amounts of free-volume are based primarily on chemical synthesis of new polymers. Here we report a simple methodology for generating free-volume based on the self-assembly of polyethylene-b-polydimethylsiloxane-b-polyethylene triblock copolymers. We have used this method to fabricate a series of membranes with identical compositions but with different amounts of free-volume. We use the term artificial free-volume to refer to the additional free-volume created by self-assembly. The effect of artificial free-volume on selective transport through the membranes was tested using butanol/water and ethanol/water mixtures due to their importance in biofuel production. We found that the introduction of artificial free-volume improves both alcohol permeability and selectivity.
Sigma-Aldrich
1,3-Butadiene solution, 15 wt. % in hexane
Sigma-Aldrich
1,3-Butadiene solution, 20 wt. % in toluene
Sigma-Aldrich
sec-Butyllithium solution, 1.4 M in cyclohexane
Sigma-Aldrich
1,3-Butadiene, ≥99%
Sigma-Aldrich
1,2-Bis(chlorodimethylsilyl)ethane, 96%
Sigma-Aldrich
Hexamethylcyclotrisiloxane, 98%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%