Merck
  • A label-free method for detecting biothiols based on poly(thymine)-templated copper nanoparticles.
A label-free method for detecting biothiols based on poly(thymine)-templated copper nanoparticles.
Biosensors & bioelectronics (2015-02-24)
Lin Zhang, Qi-Yong Cai, Jie Li, Jia Ge, Jun-Yan Wang, Zhen-Zhen Dong, Zhao-Hui Li
ABSTRACT
A novel label-free nanosensor has been developed for detecting biothiols including cysteine, glutathione and homocysteine based on poly(thymine)-templated fluorescent copper nanoparticles (CuNPs), which were controlled through thymine-Hg(II)-thymine coordination. This assay provides a simple, cost-effective, and sensitive platform for the detection of biothiols. By employing this turn-on sensor, biological thiols, such as cysteine, glutathione and homocysteine, are successfully detected at concentrations as low as 12.5nM for cysteine, 15nM for glutathione, and 20nM for homocysteine. The sensing system also exhibits high selectivity against other amino acids, and the application of the sensor for biological fluids shows that the proposed method works well for real samples.
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
MOPS