Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibition of MMP-13 prevents diet-induced obesity in mice and suppresses adipogenesis in 3T3-L1 preadipocytes.
Inhibition of MMP-13 prevents diet-induced obesity in mice and suppresses adipogenesis in 3T3-L1 preadipocytes.
Molecular biology reports (2015-02-16)
Chia-Li M Shih, Kolapo M Ajuwon
ABSTRACT
Adipose tissue remodeling by the matrix metalloproteases (MMPs) is critical for tissue hypertrophy and obesity. MMP-13 is an important protein that is highly expressed in adipose tissue but whose potential role in adipose tissue expansion is poorly characterized. We investigated the effect of pharmacological inhibition of MMP-13 with a selective inhibitor, CP-544439, on adipose tissue mass in mice on a high fat diet, and determined the effect of the inhibitor during in vitro adipocyte differentiation of 3T3-L1 cells. CP-544439 was administered for 6 weeks to mice on a high fat diet. Body adiposity and glucose tolerance was determined. Differentiating 3T3-L1 adipocytes were also treated with the inhibitor for a maximum of 8 days and adipogenesis assessed. Treatment of mice with the inhibitor resulted in reduction in body adiposity and improvement in glucose clearance. Histological examination of epididymal adipose showed reduced adipocyte hypertrophy accompanied by increased staining for collagen in the inhibitor treated mice. Treatment of differentiating 3T3-L1 cells with the inhibitor resulted in reduced adipocyte differentiation. Knockdown of MMP-13 using small interfering RNA in differentiating 3T3-L1 cells reduced adipocyte differentiation indicated by reduced expression of PPARγ. These results suggest that MMP-13 may play a major role in adipose development and its inhibition could be a potential strategy to prevent obesity.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP13