Merck
  • 68Ga- and 177Lu-Labeled PSMA I&T: Optimization of a PSMA-Targeted Theranostic Concept and First Proof-of-Concept Human Studies.
68Ga- and 177Lu-Labeled PSMA I&T: Optimization of a PSMA-Targeted Theranostic Concept and First Proof-of-Concept Human Studies.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2015-06-20)
Martina Weineisen, Margret Schottelius, Jakub Simecek, Richard P Baum, Akin Yildiz, Seval Beykan, Harshad R Kulkarni, Michael Lassmann, Ingo Klette, Matthias Eiber, Markus Schwaiger, Hans-Jürgen Wester
ABSTRACT
On the basis of the high and consistent expression of prostate-specific membrane antigen (PSMA) in metastatic prostate cancer (PC), the goal of this study was the development, preclinical evaluation, and first proof-of-concept investigation of a PSMA inhibitor for imaging and therapy (PSMA I&T) for (68)Ga-based PET and (177)Lu-based endoradiotherapeutic treatment in patients with metastatic and castration-resistant disease. PSMA I&T was synthesized in a combined solid phase and solution chemistry strategy. The PSMA affinity of (nat)Ga-/(nat)Lu-PSMA I&T was determined in a competitive binding assay using LNCaP cells. Internalization kinetics of (68)Ga- and (177)Lu-PSMA I&T were investigated using the same cell line, and biodistribution studies were performed in LNCaP tumor-bearing CD-1 nu/nu mice. Initial human PET imaging studies using (68)Ga-PSMA I&T, as well as endoradiotherapeutic treatment of 2 patients with metastatic PC using (177)Lu-PSMA I&T, were performed. PSMA I&T and its cold gallium and lutetium analog revealed nanomolar affinity toward PSMA. The DOTAGA (1,4,7,10-tetraazacyclododecane-1-(glutamic acid)-4,7,10-triacetic acid) conjugate PSMA I&T allowed fast and high-yield labeling with (68)Ga(III) and (177)Lu(III). Uptake of (68)Ga-/(177)Lu-PSMA I&T in LNCaP tumor cells is highly efficient and PSMA-specific, as demonstrated by competition studies both in vitro and in vivo. Tumor targeting and tracer kinetics in vivo were fast, with the highest uptake in tumor xenografts and kidneys (both PSMA-specific). First-in-human (68)Ga-PSMA I&T PET imaging allowed high-contrast detection of bone lesions, lymph node, and liver metastases. Endoradiotherapy with (177)Lu-PSMA I&T in 2 patients was found to be effective and safe with no detectable side effects. (68)Ga-PSMA I&T shows potential for high-contrast PET imaging of metastatic PC, whereas its (177)Lu-labeled counterpart exhibits suitable targeting and retention characteristics for successful endoradiotherapeutic treatment. Prospective studies on larger cohorts of patients are warranted and planned.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems