Merck
  • Effect of Suckling Systems on Serum Oxytocin and Cortisol Concentrations and Behavior to a Novel Object in Beef Calves.
Effect of Suckling Systems on Serum Oxytocin and Cortisol Concentrations and Behavior to a Novel Object in Beef Calves.
Asian-Australasian journal of animal sciences (2015-11-19)
Siyu Chen, Shigefumi Tanaka, Shin-Ichiro Ogura, Sanggun Roh, Shusuke Sato
ABSTRACT
We investigated differences between effects of natural- and bucket-suckling methods on basal serum oxytocin (OT) and cortisol concentrations, and the effect of OT concentration on affiliative and investigative behavior of calves to a novel object. Ten Japanese Black calves, balanced with birth order, were allocated evenly to natural-suckling (NS) and bucket suckling (BS) groups. Blood samples were collected at the ages of 1 and 2 months (1 week after weaning) calves, and serum OT and cortisol concentrations were measured using enzyme-linked immunosorbent assay and enzymeimmunoassay tests, respectively. Each calf at the age of 2 months (2 weeks after weaning) was released into an open-field with a calf decoy, and its investigative and affiliative behaviors were recorded for 20 minutes. In 1-month-old calves, the basal serum OT concentration (25.5±4.9 [mean±standard deviation, pg/mL]) of NS was significantly higher than that of BS (16.9±6.7) (p<0.05), whereas the basal cortisol concentration (5.8±2.5 [mean±standard deviation, ng/mL]) of NS was significantly lower than that in BS (10.0±2.8) (p<0.05). Additionally, a negative correlation was noted between serum OT and cortisol concentrations in 1-month-old calves (p = 0.06). Further, the higher serum OT concentration the calves had at 1 month old, the more investigative the calves were at 2 months old but not affiliative in the open-field with a calf decoy. Thus, we concluded that the natural suckling method from a dam elevates the basal serum OT concentration in calves, and high serum OT concentrations induce investigative behavior and attenuate cortisol concentrations.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)