Merck
  • Controlling the Balance of Fibroblast Proliferation and Differentiation: Impact of Thy-1.
Controlling the Balance of Fibroblast Proliferation and Differentiation: Impact of Thy-1.
The Journal of investigative dermatology (2015-03-05)
Manuela Schmidt, Danny Gutknecht, Jan C Simon, Jan-Niklas Schulz, Beate Eckes, Ulf Anderegg, Anja Saalbach
ABSTRACT
Fibroblasts provide matrix and regulatory mediators to the microenvironment and thereby contribute to maintenance of tissue homeostasis, wound healing, and tumor progression. In the present study, we addressed the functional relevance of Thy-1 for fibroblast functions in vitro and in vivo. Using fibroblasts from Thy-1(-/-) and wild-type mice, recombinant expression of Thy-1, and analysis of the interaction of fibroblasts with immobilized Thy-1, we showed that Thy-1 has a crucial role in the control of cell growth by suppressing proliferation and promoting apoptosis and differentiation of dermal fibroblasts. Function-blocking studies revealed that Thy-1 mediates the control of apoptosis and proliferation via modulation of β3 integrin function. Interestingly, Thy-1-mediated growth control appears to be a more general mechanism because it also regulates growth of tumor cells of different origin dependent on their β3 integrin expression. In summary, our findings point to an important role of Thy-1 in controlling the balance between proliferation and differentiation in dermal fibroblasts.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C