Merck
  • A Hyaluronic Acid-Conjugated Gadolinium Hepatocyte-Specific T1 Contrast Agent for Liver Magnetic Resonance Imaging.
A Hyaluronic Acid-Conjugated Gadolinium Hepatocyte-Specific T1 Contrast Agent for Liver Magnetic Resonance Imaging.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2015-01-22)
MyeongJu Moon, Reju George Thomas, Seon-U Heo, Myong-Suk Park, Woo Kyun Bae, Suk Hee Heo, Nam Yeol Yim, Yong Yeon Jeong
ABSTRACT
In this study, we synthesized hyaluronic acid-conjugated gadolinium (HA-diethylene triamine pentaacetic acid (DTPA)-Gd) and evaluated as hepatocyte-specific magnetic resonance imaging (MRI) contrast agent for the diagnosis of hepatic metastasis. We conducted Fourier transform (FT)-IR analysis to determine the conjugation of HA and DTPA and performed cell viability assays using NIH3T3 and FL83B cell lines. We also conducted T1-weighted MRI of HA-DTPA-Gd and gadoxetic acid to compare the paramagnetic properties of both. HA-DTPA-Gd had a higher efficiency in liver MRI compared with the commercially available liver-specific contrast agent (p < 0.001). HA-DTPA-Gd, which possessed a higher T1 relaxivity, showed excellent capability for the diagnosis of hepatic metastasis through an in vivo MRI study in comparison with gadoxetic acid (p < 0.001). Based on this study, we believe that HA-DTPA-Gd has promising potential for use as a contrast agent for liver MRI of hepatic metastases.
Sigma-Aldrich
Gadolinium(III) chloride hydrate, 99.99% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Diethylenetriaminepentaacetic dianhydride, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Gadolinium, wire reel, 25mm, diameter 0.5mm, hard, 99.9%
Gadolinium, rod, 100mm, diameter 6.35mm, cast, 99%
Gadolinium, wire reel, 100mm, diameter 0.5mm, hard, 99.9%
Gadolinium, rod, 100mm, diameter 12.5mm, cast, 99%
Gadolinium, rod, 100mm, diameter 3.0mm, cast, 99%
Gadolinium, rod, 50mm, diameter 12.5mm, cast, 99%
Gadolinium, rod, 50mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Gadolinium, rod, 48mm, diameter 3.0mm, cast, 99%
Gadolinium, wire reel, 50mm, diameter 0.5mm, hard, 99.9%