Merck
  • TSPAN8 promotes gastric cancer growth and metastasis via ERK MAPK pathway.
TSPAN8 promotes gastric cancer growth and metastasis via ERK MAPK pathway.
International journal of clinical and experimental medicine (2015-08-27)
Lunshou Wei, Yan Li, Zhimin Suo
ABSTRACT
This study was designed to investigate the effects of Tetraspanin 8 (TSPAN8) overexpression and TSPAN8 suppression on gastric cancer cell proliferation and invasion. Furthermore, whether extracellular-signal regulated kinase (ERK) mitogen-activated protein kinase (MAPK) pathway was involved in TSPAN8's function on gastric cancer cells was examined. The expression of TSPAN8 in human gastric cancer tissues and gastric cancer cell lines was detected using real-time PCR and western blot analysis. TSPAN8-pcDNA3.1 plasmid or TSPAN8 siRNA was transfected into the gastric cancer cell lines to overexpress or suppress TSPAN8. Cells were treated with U0126 to inhibit ERK MAPK pathway. Cell proliferation and invasion were assessed by MTT and transwell-matrigel assay. TSPAN8 was overexpressed in human gastric cancer tissues and gastric cancer cell lines compared with the normal. TSPAN8 overexpression promoted cell proliferation and invasion, while TSPAN8 suppression inhibited cell proliferation and invasion. TSPAN8 could activate the ERK MAPK pathway in gastric cancer cells, and MEK-ERK inhibition reversed the effects of TSPAN8 overexpression on cell proliferation and invasion. This study firstly demonstrated that TSPAN8 promotes gastric cancer cell growth and metastasis at least partially through the activation of ERK MAPK pathway. These findings provided a novel molecular basis for the understanding and treatment of gastric cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tspan8
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TSPAN8