Merck
  • Synthesis, antiproliferative and antifungal activities of 1,2,3-triazole-substituted carnosic Acid and carnosol derivatives.
Synthesis, antiproliferative and antifungal activities of 1,2,3-triazole-substituted carnosic Acid and carnosol derivatives.
Molecules (Basel, Switzerland) (2015-05-27)
Mariano Walter Pertino, Cristina Theoduloz, Estefania Butassi, Susana Zacchino, Guillermo Schmeda-Hirschmann
ABSTRACT
Abietane diterpenes exhibit an array of interesting biological activities, which have generated significant interest among the pharmacological community. Starting from the abietane diterpenes carnosic acid and carnosol, twenty four new triazole derivatives were synthesized using click chemistry. The compounds differ in the length of the linker and the substituent on the triazole moiety. The compounds were assessed as antiproliferative and antifungal agents. The antiproliferative activity was determined on normal lung fibroblasts (MRC-5), gastric epithelial adenocarcinoma (AGS), lung cancer (SK-MES-1) and bladder carcinoma (J82) cells while the antifungal activity was assessed against Candida albicans ATCC 10231 and Cryptococcus neoformans ATCC 32264. The carnosic acid γ-lactone derivatives 1-3 were the most active antiproliferative compounds of the series, with IC50 values in the range of 43.4-46.9 μM and 39.2-48.9 μM for MRC-5 and AGS cells, respectively. Regarding antifungal activity, C. neoformans was the most sensitive fungus, with nine compounds inhibiting more than 50% of its fungal growth at concentrations ≤250 µg∙mL-1. Compound 22, possessing a p-Br-benzyl substituent on the triazole ring, showed the best activity (91% growth inhibition) at 250 µg∙mL-1 In turn, six compounds inhibited 50% C. albicans growth at concentrations lower than 250 µg∙mL-1.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, Vetec, reagent grade, ≥99.5% (HPLC)