Merck
  • Enhanced osteogenesis on titanium implants by UVB photofunctionalization of hydrothermally grown TiO₂ coatings.
Enhanced osteogenesis on titanium implants by UVB photofunctionalization of hydrothermally grown TiO₂ coatings.
Journal of biomaterials applications (2015-01-31)
Martina Lorenzetti, Olga Dakischew, Katja Trinkaus, Katrin Susanne Lips, Reinhard Schnettler, Spomenka Kobe, Saša Novak
ABSTRACT
Even though Ti-based implants are the most used materials for hard tissue replacement, they may present lack of osseointegration on the long term, due to their inertness. Hydrothermal treatment (HT) is a useful technique for the synthesis of firmly attached, highly crystalline coatings made of anatase titanium dioxide (TiO2), providing favorable nanoroughness and higher exposed surface area, as well as greater hydrophilicity, compared to the native amorphous oxide on pristine titanium. The hydrophilicity drops even more by photofunctionalization of the nanostructured TiO2-anatase coatings under UV light. Human mesenchymal stem cells exhibited a good response to the combination of the positive surface characteristics, especially in respect to the UVB pre-irradiation. The results showed that the cells were not harmed in terms of viability; even more, they were encouraged to differentiate in osteoblasts and to become osteogenically active, as confirmed by the calcium ion uptake and the formation of well-mineralized, bone-like nodule structures. In addition, the enrichment of hydroxyl groups on the HT-surfaces by UVB photofunctionalization accelerated the cell differentiation process and greatly improved the osteogenesis in comparison with the nonirradiated samples. The optimal surface characteristics of the HT-anatase coatings as well as the high potentiality of the photo-induced hydrophilicity, which was reached during a relatively short pre-irradiation time (5 h) with UVB light, can be correlated with better osseointegration ability in vivo; among the samples, the superior biological behavior of the roughest and most hydrophilic HT coating makes it a good candidate for further studies and applications.
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trimethylphenylammonium hydroxide solution, ~25% in H2O (1.68 M)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Phenylacetic acid, 99%