Merck
  • Histone demethylase Utx regulates differentiation and mineralization in osteoblasts.
Histone demethylase Utx regulates differentiation and mineralization in osteoblasts.
Journal of cellular biochemistry (2015-04-29)
Di Yang, Hirohiko Okamura, Jumpei Teramachi, Tatsuji Haneji
ABSTRACT
Alteration of methylation status of lysine 27 on histone H3 (H3K27) associates with dramatic changes in gene expression in response to various differentiation signals. Demethylation of H3K27 is controlled by specific histone demethylases including ubiquitously transcribed tetratricopeptide repeat X chromosome (Utx). However, the role of Utx in osteoblast differentiation remains unknown. In this study, we examined whether Utx should be involved in osteoblast differentiation. Expression of Utx increased during osteoblast differentiation in MC3T3-E1 cells and primary osteoblasts. GSK-J1, a potent inhibitor of H3K27 demethylase, increased the levels of trimethylated H3K27 (H3K27me3) and decreased the expressions of Runx2 and Osterix and ALP activity in MC3T3-E1 cells. Stable knockdown of Utx by shRNA attenuated osteoblast differentiation and decreased ALP activity, calcium content, and bone-related gene expressions. Silencing of Utx increased the level of H3K27me3 on the promoter regions of Runx2 and Osterix and decreased the promoter activities of Runx2 and Osterix. Taken together, our present results propose that Utx plays important roles in osteoblast differentiation by controlling the expressions of Runx2 and Osterix.
Sigma-Aldrich
Lithium-7Li2 carbonate, ≥99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Lithium carbonate, 99.999% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting human KDM6A (1)
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Cacodylic acid, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O