Merck
  • Effects of cytoplasm and reactant polarities on acid-catalyzed lipid transesterification in wet microalgal cells subjected to microwave irradiation.
Effects of cytoplasm and reactant polarities on acid-catalyzed lipid transesterification in wet microalgal cells subjected to microwave irradiation.
Bioresource technology (2015-11-13)
Rui Huang, Jun Cheng, Yi Qiu, Tao Li, Junhu Zhou, Kefa Cen
ABSTRACT
The polarities of the cytoplasm and reactants were measured through dielectric spectroscopy, contact angle test, NMR, and FTIR to investigate the mechanisms underlying acid-catalyzed lipid transesterification in wet microalgal cells subjected to microwave irradiation. Organics with apolar functional groups in the cytoplasm decreased the contact angle of methanol against triglyceride by 13.92°, which subsequently increased transesterification efficiency by 2.4 times. The microalgal biomass, given its higher hydrophilicity index of 1.96 than lipids, was more accessible to hydrophilic alcohols, which subsequently promoted transesterification. Water in the cytoplasm promoted the dielectric constant of methanol and increased the contact angle of methanol against triglyceride by 20.51°, which subsequently decreased transesterification efficiency by 72.6%. The inhibitory effect of water on transesterification weakened with the prolonged carbon lengths of the alcohols because of decreased polarity. Microwave decreased the electric constants of alcohols and reduced the polarity difference between alcohols and lipids, thereby improving transesterification efficiency.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%