Merck
  • Chemical and physical properties of carbonated hydroxyapatite affect breast cancer cell behavior.
Chemical and physical properties of carbonated hydroxyapatite affect breast cancer cell behavior.
Acta biomaterialia (2015-06-15)
Siyoung Choi, Scott Coonrod, Lara Estroff, Claudia Fischbach
ABSTRACT
Breast microcalcifications are routinely explored for mammographic detection of breast cancer and are primarily composed of non-stoichiometric hydroxyapatite (Ca10-x(PO4)6-x(CO3)x(OH)2-x) (HA). Interestingly, HA morphology and carbonate substitution vary in malignant vs. benign lesions. However, whether or not these changes (i) are functionally linked and (ii) impact malignancy remains unclear due in part to lack of model systems that permit evaluating these possibilities. Here, we have adapted a 96 well-based mineralized culture platform to investigate breast cancer cell behavior in response to systematic changes in the chemical and physical properties of HA. By adjusting the carbonate content of the simulated body fluid (SBF) solutions used during growth, we can control the morphology and carbonate substitution of the deposited HA. Our results suggest that both the combined and individual effects of these differences alter breast cancer cell growth and secretion of tumorigenic interleukin-8 (IL-8). Consequently, changes in both HA carbonate incorporation and morphology impact the behavior of breast cancer cells. Collectively, our data underline the importance of biomineralized culture platforms to evaluate the functional contribution of HA material properties to the pathogenesis of breast cancer. Breast microcalcifications are small mineral deposits primarily composed of hydroxyapatite (HA). HA physicochemical properties have been of considerable interest, as these are often altered during breast cancer progression and linked to malignancy. However, the functional relationship between these changes and malignancy remains unclear due in part to lack of model systems. Here, we have adapted a previously developed a 96 well-based culture platform to evaluate breast cancer cell behavior in response to systematic changes in HA properties. Our results demonstrate that changes in HA morphology and carbonate content influence breast cancer cell growth and interleukin-8 secretion, and suggest that characterizing the effect of HA properties on breast cancer cells may improve our understanding of breast cancer development and progression.
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon