Merck
  • Measurement of low amounts of amorphous content in hydrophobic active pharmaceutical ingredients with dynamic organic vapor sorption.
Measurement of low amounts of amorphous content in hydrophobic active pharmaceutical ingredients with dynamic organic vapor sorption.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-03-18)
Thorsten Müller, Jörg Schiewe, Rüdiger Smal, Claudius Weiler, Markus Wolkenhauer, Hartwig Steckel
ABSTRACT
Today, a variety of devices for dry powder inhalers (DPIs) is available and many different formulations for optimized deposition in the lung are developed. However, during the production of powder inhalers, processing steps may induce changes to both, the carrier and active pharmaceutical ingredients (APIs). It is well known that standard pharmaceutical operations may lead to structural changes, crystal defects and amorphous regions. Especially operations such as milling, blending and even sieving generate these effects. These disorders may induce re-crystallization and particle size changes post-production which have a huge influence on drug delivery and product stability. In this study, pilot tests with a polar solvent (water) and hydrophilic drug (Salbutamol sulfate) were performed to receive a first impression on further possible implementation of hydrophobic samples with organic solvents. Thereafter, a reliable method for the accurate detection of low amounts of amorphous content is described up to a limit of quantification (LOQ) of 0.5% for a hydrophobic model API (Ciclesonide). The organic vapor sorption method which is a gravimetric method quantifies exactly these low amounts of amorphous content in the hydrophobic powder once the suitable solvent (isopropanol), the correct p/p0 value (0.1) and the exact temperature (25°C) have been found. Afterward it was possible to quantitate low amorphous amounts in jet-milled powders (0.5-17.0%). In summary, the data of the study led to a clearer understanding in what quantity amorphous parts were generated in single production steps and how variable these parts behave to fully crystalline material. Nevertheless it showed how difficult it was to re-crystallize hydrophobic material with water vapor over a short period. For the individual samples it was possible to determine the single humidity at which the material starts to re-crystallize, the behavior against different nonpolar solvents and the calculation of the reduction of the glass transition temperature (Tg) according to the Gordon-Taylor equation.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate