Merck
  • Clonal restriction and predominance of regulatory T cells in coronary thrombi of patients with acute coronary syndromes.
Clonal restriction and predominance of regulatory T cells in coronary thrombi of patients with acute coronary syndromes.
European heart journal (2014-01-15)
Roland Klingenberg, Chad E Brokopp, Audrey Grivès, Anaïs Courtier, Milosz Jaguszewski, Nicolas Pasqual, Eugenia Vlaskou Badra, Anika Lewandowski, Oliver Gaemperli, Simon P Hoerstrup, Willibald Maier, Ulf Landmesser, Thomas F Lüscher, Christian M Matter
ABSTRACT
Regulatory T cells (Treg) exert anti-inflammatory and atheroprotective effects in experimental atherosclerosis. Treg can be induced against specific antigens using immunization strategies associated with clonal restriction. No data exist on Treg in combination with clonal restriction of T cells in patients with acute coronary syndromes (ACS). Among T cell subsets characterized by flow cytometry, Treg (CD4(+) CD25(+) CD127(low)) were twice as frequent in coronary thrombi compared with peripheral blood. Treg prevailed among T cell subsets identified in coronary thrombi. To evaluate clonal restriction, genomic DNA was extracted from coronary thrombi and peripheral blood in order to evaluate T cell receptor (TCR) β chain diversity by means of Multi-N-plex PCR using a primer specific for all TCR β V gene segments and another primer specific for TCR β J gene segments. T cell receptor diversity was reduced in thrombi compared with peripheral blood (intra-individual comparisons in 16 patients) with 8 gene rearrangements in the TCR common in at least 6 out of 16 analysed coronary thrombi. Compared with age-matched healthy controls (n = 16), TCR diversity was also reduced in peripheral blood of patients with ACS; these findings were independent of peripheral T cell numbers. We provide novel evidence for a perturbed T cell compartment characterized by clonal restriction in peripheral blood and coronary thrombi from patients with ACS. Our findings warrant further studies on Treg as novel therapeutic targets aimed at enhancing this anti-inflammatory component of adaptive immunity in human atherothrombosis.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%