- Application of Mannich bases to the synthesis of hydroxymethylated isoflavonoids as potential antineoplastic agents.
Application of Mannich bases to the synthesis of hydroxymethylated isoflavonoids as potential antineoplastic agents.
The regiospecific Mannich aminomethylation of 7-hydroxyisoflavonoids using bis(N,N-dimethylamino)methane afforded C-8 substituted N,N-dimethylaminomethyl adducts, and the regioselective aminomethylation of 5-hydroxy-7-methoxyisoflavonoids afforded predominantly the C-6 substituted N,N-dimethylaminomethyl adducts. Acetylation of these C-6 or C-8 Mannich bases with potassium acetate in acetic anhydride provided access to the corresponding acetoxymethyl derivatives that were subsequently converted to hydroxymethyl- and methoxymethyl-substituted 5-hydroxy- or 7-hydroxyisoflavonoids related to naturally occurring flavonoids. The C-8 acetoxymethyl, hydroxymethyl or methoxymethyl-substituted isoflavonoids possessed promising inhibitory potency in the low micromolar range in a prostate cancer PC-3 cell proliferation assay.