Merck
  • Sterol liganding of OSBP-related proteins (ORPs) regulates the subcellular distribution of ORP-VAPA complexes and their impacts on organelle structure.
Sterol liganding of OSBP-related proteins (ORPs) regulates the subcellular distribution of ORP-VAPA complexes and their impacts on organelle structure.
Steroids (2015-02-15)
Henriikka Kentala, Simon G Pfisterer, Vesa M Olkkonen, Marion Weber-Boyvat
ABSTRACT
Oxysterol-binding protein (OSBP) and its homologues (ORPs) are lipid-binding/transfer proteins with affinity for oxysterols, cholesterol and glycerophospholipids. In addition to a ligand-binding domain, a majority of the ORPs carry a pleckstrin homology domain that targets organelle membranes via phosphoinositides, and a motif targeting the endoplasmic reticulum (ER) via VAMP-associated proteins (VAPs). We employed here Bimolecular Fluorescence Complementation (BiFC) to systematically assess the effects of sterol manipulation of HuH7 cells on complexes of established sterol-binding ORPs with their ER receptor, VAMP-associated protein A (VAPA). Depletion of cellular cholesterol with lipoprotein-deficient medium and Mevastatin caused concentration of OSBP-VAPA complexes and Golgi complex markers at a juxtanuclear position, an effect reversed by low-density lipoprotein treatment. A similar redistribution of OSBP-VAPA but not of sterol-binding deficient mutant OSBP(ΔELSK)-VAPA, occurred upon treatment with the high-affinity ligand, 25-hydroxycholesterol (25OHC), which reduced total and free cholesterol. ORP2-VAPA complexes, which localize in untreated cells at blob-like ER structures with associated lipid droplets, were redistributed upon treatment with the ORP2 ligand 22(R)OHC to a diffuse cytoplasmic/ER pattern and the plasma membrane. Analogously, distribution of ORP4L-VAPA complexes between the plasma membrane and vimentin intermediate filament associated compartments was modified by statin or 25OHC treatment. The treatments resulted in loss of vimentin co-localization, and sterol-binding deficient ORP4L(ΔELSR)-VAPA localized predominantly to the plasma membrane. In conclusion, treatment with statin or oxysterol ligands modify the subcellular targeting of ORP-VAPA complexes, consistent with the notion that this machinery controls lipid homeostasis and signaling at organelle interfaces.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Octane, anhydrous, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining