Merck
  • Characterization of modified proteins in plasma from a subtype of schizophrenia based on carbonyl stress: Protein carbonyl is a possible biomarker of psychiatric disorders.
Characterization of modified proteins in plasma from a subtype of schizophrenia based on carbonyl stress: Protein carbonyl is a possible biomarker of psychiatric disorders.
Biochemical and biophysical research communications (2015-10-04)
Shin Koike, Tasuku Kayama, Makoto Arai, Yasue Horiuchi, Akiko Kobori, Mitsuhiro Miyashita, Masanari Itokawa, Yuki Ogasawara
ABSTRACT
Although it's well known that protein carbonyl (PCO) and advanced glycation end-products (AGEs) levels are elevated in plasma from patients with renal dysfunction, we recently identified patients who had no renal dysfunction but possessed high levels of plasma pentosidine (PEN), which is an AGEs, and low vitamin B6 levels in serum. In this study, we investigated the status of carbonyl stress to characterize the subtype of schizophrenia. When plasma samples were subjected to Western blot analysis for various AGEs, clear differences were only observed with the anti-PEN antibody in the plasma from schizophrenic patients. Moreover, we determined the formation of protein carbonyl (PCO), a typical indicator of carbonyl stress, occurred prior to the accumulation of PEN in the plasma of schizophrenic patients. PCO levels in the plasma from schizophrenic patients were significantly higher than that from healthy subjects. Western blots analysis clearly showed that albumin and IgG were markedly carbonylated in the plasma of some patients. Thus, PCOs may be a novel marker of carbonyl stress-type schizophrenia in addition to albumin containing PEN structure.
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O