Merck
  • Characterisation of porcine dermis scaffolds decellularised using a novel non-enzymatic method for biomedical applications.
Characterisation of porcine dermis scaffolds decellularised using a novel non-enzymatic method for biomedical applications.
Journal of biomaterials applications (2015-04-10)
K V Greco, L Francis, M Somasundaram, G Greco, Nicholas R English, Judith A Roether, Aldo R Boccaccini, P Sibbons, T Ansari
ABSTRACT
Off-the-shelf availability of tissue-engineered skin constructs, tailored by different combinations of reagents to produce a highly preserved biological matrix is often the only means to help patients suffering skin damage. This study assessed the effect of five different decellularisation methods on porcine dermal scaffolds with regard to matrix composition, biomechanical strength, and cytotoxicity using an in vitro biocompatibility assay. Results demonstrated that four out of the five tested decellularisation protocols were efficient in producing acellular scaffolds. Nevertheless, decellularisation method using osmotic shock without enzymatic digestion showed to be efficient not only in removing cellular material and debris from dermal scaffolds but was also beneficial in the preservation of extracellular matrix components (glycosaminoglycans and collagen). Histological assessment revealed that the dermal architecture of coarse collagen bundles was preserved. Examinations by scanning electron microscopy and transmission electron microscopy showed that the arrangement and ultrastructure of collagen fibrils in the scaffolds were retained following non-enzymatic method of decellularisation and also after collagen crosslinking using genipin. Moreover, this decellularised scaffold was not only shown to be biologically compatible when co-cultured with bone marrow-derived mesenchymal stem cells and fibroblasts, but also stimulated the cells to release trophic factors essential for tissue regeneration.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)