Merck
  • Identification of a Candidate Gene Panel for the Early Diagnosis of Prostate Cancer.
Identification of a Candidate Gene Panel for the Early Diagnosis of Prostate Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-03-20)
Gisele H J M Leyten, Daphne Hessels, Frank P Smit, Sander A Jannink, Hans de Jong, Willem J G Melchers, Erik B Cornel, Theo M de Reijke, Henk Vergunst, Paul Kil, Ben C Knipscheer, Christina A Hulsbergen-van de Kaa, Peter F A Mulders, Inge M van Oort, Jack A Schalken
ABSTRACT
Serum PSA (sPSA) testing has led to the identification of patients with indolent prostate cancer, and inevitably overtreatment has become a concern. Progensa PCA3 urine testing was shown to improve the diagnosis of prostate cancer, but its diagnostic value for aggressive prostate cancer is limited. Therefore, urinary biomarkers that can be used for prediction of Gleason score ≥7 prostate cancer in biopsies are urgently needed. Using gene expression profiling data, 39 prostate cancer biomarkers were identified. After quantitative PCR analysis on tissue specimens and urinary sediments, eight promising biomarkers for the urinary detection of prostate cancer were selected (ONECUT2, HOXC4, HOXC6, DLX1, TDRD1, NKAIN1, MS4A8B, PPFIA2). The hypothesis that biomarker combinations improve the diagnostic value for aggressive prostate cancer was tested on 358 urinary sediments of an intention-to-treat cohort. A urinary three-gene panel (HOXC6, TDRD1, and DLX1) had higher accuracy [area under the curve (AUC), 0.77; 95% confidence interval (CI), 0.71-0.83] to predict Gleason score ≥7 prostate cancer in biopsies compared with Progensa PCA3 (AUC, 0.68; 95% CI, 0.62-0.75) or sPSA (AUC, 0.72; 95% CI, 0.65-0.78). Combining the three-gene panel with sPSA further improved the predictive accuracy (AUC, 0.81; 95% CI, 0.75-0.86). The accuracy of the three-gene predictive model was maintained in subgroups with low sPSA concentrations. The urinary three-gene panel (HOXC6, TDRD1, and DLX1) represents a promising tool to identify patients with aggressive prostate cancer, also in those with low sPSA values. The combination of the urinary three-gene panel with sPSA bears great potential for the early diagnosis of patients with clinically significant prostate cancer.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis