Merck
  • Sorafenib and DE605, a novel c-Met inhibitor, synergistically suppress hepatocellular carcinoma.
Sorafenib and DE605, a novel c-Met inhibitor, synergistically suppress hepatocellular carcinoma.
Oncotarget (2015-04-22)
Xiufeng Jiang, Kang Feng, Ye Zhang, Zengyao Li, Fan Zhou, Huiqiang Dou, Tong Wang
ABSTRACT
Sorafenib, an oral multikinase inhibitor of Raf, VEGF and PDGF receptor signaling is approved for advanced hepatocellular carcinoma (HCC). One strategy to improve HCC therapy is to combine agents that target key signaling pathways. Aberrant mesenchymal-epithelial transition factor (c-Met) activation is associated with a variety of human malignancies and therefore represents a target for therapy. In this study, we investigated a novel c-Met inhibitor, DE605, together with sorafenib in hepatocellular carcinoma cells in vitro and in vivo. DE605 and sorafenib synergistically induced apoptosis in hepatocellular carcinoma cells. Mechanistically, DE605 activated the FGFR3/Erk pathway, which in turn was inhibited by sorafenib, resulting in synergism. Finally, DE605 and sorafenib significantly inhibited growth of PLC/PRF/5 hepatocellular carcinoma tumor xenografts in athymic nude mice. Importantly, no obvious weight loss (toxicity) was detected. Thus in combination, DE605 and sorafenib target complementary anti-apoptotic pathways and synergistically suppress HCC, providing the rationale for clinical studies with this novel combination.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR3 (1)
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK1
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR3 (2)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Z-VAD-FMK, solid
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%