Merck
  • Calcium-induced calcium release and gap junctions mediate large-scale calcium waves in olfactory ensheathing cells in situ.
Calcium-induced calcium release and gap junctions mediate large-scale calcium waves in olfactory ensheathing cells in situ.
Cell calcium (2015-06-21)
Maren Stavermann, Patrick Meuth, Michael Doengi, Anne Thyssen, Joachim W Deitmer, Christian Lohr
ABSTRACT
Olfactory ensheathing cells (OECs) are a specialised type of glial cells, supporting axon growth and guidance during development and regeneration of the olfactory nerve and the nerve layer of the olfactory bulb. We measured calcium signalling in OECs in olfactory bulb in-toto preparations using confocal and epifluorescence microscopy and the calcium indicator Fluo-4. We identified two subpopulations of olfactory bulb OECs: OECs in the outer sublamina of the nerve layer responded to purinergic neurotransmitters such as adenosine triphosphate with calcium transients, while OECs in the inner sublamina of the nerve layer did not respond to neurotransmitters. However, the latter generated spontaneous calcium waves that covered hundreds of cells. These calcium waves persisted in the presence of tetrodotoxin and in calcium-free saline, but were abolished after calcium store depletion with cyclopiazonic acid or inositol trisphosphate receptor blockage with 2-APB. Calcium waves could be triggered by laser photolysis of caged inositol trisphosphate. Blocking purinoceptors with PPADS had no effect on calcium wave propagation, whereas blocking gap junctions with carbenoxolone or meclofenamic acid entirely suppressed calcium waves. Increasing calcium buffer capacity in OECs with NP-EGTA ("caged" Ca(2+)) prevented calcium wave generation, and laser photolysis of NP-EGTA in a small group of OECs resulted in a calcium increase in the irradiated cells followed by a calcium wave. We conclude that calcium waves in OECs can be initiated by calcium-induced calcium release via InsP3 receptors and propagate through gap junctions, while purinergic signalling is not involved.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sulforhodamine 101, Dye content ~95 %
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%