Merck
Biological responses to flexion/extension in spinal segments ex-vivo.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2015-04-14)
Robert A Hartman, Takashi Yurube, Kevin Ngo, Nicolas E Merzlak, Richard E Debski, Bryan N Brown, James D Kang, Gwendolyn A Sowa
ABSTRACT
Mechanical loading is a salient factor in the progression of spinal disorders that contribute to back pain. Biological responses to loading modes like flexion/extension (F/E) in relevant spinal tissues remain unstudied. A novel, multi-axial experimental system was developed to subject viable functional spinal units (FSUs) to complex, in-situ loading. The objective was to determine biological effects of F/E in multiple spinal tissues-annulus fibrosus, nucleus pulposus, facet cartilage, and ligamentum flavum. Rabbit lumbar FSUs were mounted in a bioreactor within a robotic testing system. FSUs underwent small (0.17/0.05 Nm) and large (0.5/0.15 Nm) range-of-motion F/E for 1 or 2 h of cycling. Outcomes in each tissue, compared to unloaded FSUs, included (i) relative mRNA expression of catabolic (MMP-1, 3 and ADAMTS-5), pro-inflammatory (COX-2), and anabolic (ACAN) genes and (ii) immunoblotting of aggrecan degradation. Total energy applied to FSUs increased in groups subject to large range-of-motion and 2-h cycling, and moment relaxation was higher with large range-of-motion. F/E significantly modulated MMP1,-3 and COX-2 in facet cartilage and MMP-3 and ACAN in annulus fibrosus. Large range-of-motion loading increased MMP-mediated aggrecan fragmentation in annulus fibrosus. Biological responses to complex loading ex vivo showed variation among spinal tissues that reflect tissue structure and mechanical loading in F/E.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
3-Methylbutanol, for molecular biology, BioReagent, ≥98.5%