Merck
  • Application Of Phenol/Amine Copolymerized Film Modified Magnesium Alloys: Anticorrosion And Surface Biofunctionalization.
Application Of Phenol/Amine Copolymerized Film Modified Magnesium Alloys: Anticorrosion And Surface Biofunctionalization.
ACS applied materials & interfaces (2015-10-20)
Si Chen, Jiang Zhang, Yingqi Chen, Sheng Zhao, Meiyun Chen, Xin Li, Manfred F Maitz, Jin Wang, Nan Huang
ABSTRACT
Magnesium metal as degradable metallic material is one of the most researched areas, but its rapid degradation rate restricts its development. The current anticorrosion surface modification methods require expensive equipment and complicated operation processes and cannot continue to introduce biofunction on modified surface. In this study, the GAHD conversion coatings were fabricated on the surface of magnesium alloys (MZM) by incubating in the mixture solution of gallic acid (GA) and hexamethylenediamine (HD) to decrease the corrosion rate and provide primary amines (-NH2), carboxyl (-COOH), and quinone groups, which is supposed to introduce biomolecules on MZM. Chemical structures of the MZM-GAHD and MZM-HEP-GAHD were explored by analyzing the results of FTIR and XPS comprehensively. Furthermore, it was proved that the heparin (HEP) molecules were successfully immobilized on MZM-GAHD surface through carbodiimide method. The evaluation of platelet adhesion and clotting time test showed that MZM-HEP-GAHD had higher anticoagulation than MZM-GAHD. Through electrochemical detection (polarization curves and electrochemical impedance spectroscopy Nyquist spectrum) and immersion test (Mg(2+) concentration and weight loss), it was proved that compared to MZM, both the MZM-GAHD and MZM-HEP-GAHD significantly improved the corrosion resistance. Finally, in vivo experimentation indicated that mass loss had no significant difference between MZM-1:1, MZM-HEP-1:1, and MZM. However, the trend still suggested that MZM-1:1 and MZM-HEP-1:1 possessed corrosion resistance property.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%