Merck
  • Four lanostane-type triterpenes from the fruiting bodies of mushroom Laetiporus sulphureus var. miniatus.
Four lanostane-type triterpenes from the fruiting bodies of mushroom Laetiporus sulphureus var. miniatus.
Journal of Asian natural products research (2015-04-29)
Xia Yin, Zheng-Hui Li, Yan Li, Tao Feng, Ji-Kai Liu
ABSTRACT
Two new 3,4-seco-lanostane-type triterpenes, named as 15α-hydroxy-3,4-seco-lanosta-4(28),8,24-triene-3,21-dioic acid (1), 5α-hydroxy-3,4-seco-lanosta-4(28),8,24-triene-3,21-dioic acid 3-methyl ester (2), and one new lanostane triterpene 15α-acetoxylhydroxytrametenolic acid (3) together with a known one versisponic acid D (4) were isolated from the fruiting bodies of Laetiporus sulphureus var. miniatus. Their structures were determined on the basis of extensive spectroscopic methods and comparison with reported data. All four compounds were evaluated for their cytotoxicities against five human cancer cell lines; however, none exhibited inhibitory effects.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)