Merck
  • Behavioural response of female Culex pipiens pallens to common host plant volatiles and synthetic blends.
Behavioural response of female Culex pipiens pallens to common host plant volatiles and synthetic blends.
Parasites & vectors (2015-11-19)
Bao-Ting Yu, Yan-Mei Ding, Jian-Chu Mo
ABSTRACT
Most mosquito species need to obtain sugar from host plants. Little is known about the chemical cues that Culex pipiens pallens use during their orientation to nectar host plants. In this study, we investigated the behavioural responses of female Cx. pipiens pallens to common floral scent compounds and their blends. Behavioural responses of female Cx. pipiens pallens to 18 individual compounds at different concentrations were determined in the olfactometer bioassays. A synthetic blend composed of behaviourally active compounds was formulated, and its attractiveness to mosquitoes was tested. Several most attractive compounds constituted a reduced blend, and its attractiveness was tested against the solvent and the full blend, respectively. Mosquito response in the olfactometer was analyzed by comparing the percentages of mosquitoes caught in the two arms by χ(2) test (observed versus expected). Fifteen of the 18 compounds were attractive to female Cx. pipiens pallens in the dose-dependent bioassays, with the exception of β-pinene, acetophenone and nonanal. (68.00 ± 2.49) % mosquitoes responded to the full blend composed of these 15 compounds on their optimal doses when tested against the solvent, with the preference index at 46.11 ± 3.57. Six individual compounds whose preference indices were over 40 constituted the reduced blend, and it attracted (68.00 ± 1.33) % mosquitoes when tested against the solvent while its preference index was 42.00 ± 3.54. When tested against the full blend simultaneously in the olfactometer, the reduced blend could attract (45.00 ± 2.69) % of released mosquitoes, which was as attractive as the full blend. Our results demonstrate that female Cx. pipiens pallens is differentially attracted by a variety of compounds at different concentrations. Alteration of the concentration strongly affects the attractiveness of the synthetic blend. Several floral scent volatiles might be the universal olfactory cues for various mosquito species to locate their nectar host plants, which could be potentially used in trapping devices for surveillance and control of them.
Sigma-Aldrich
Linalool oxide, natural, ≥95%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Linalool oxide, mixture of isomers, ≥97.0% (GC)
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG