Merck
  • Biomedical Activity and Related Volatile Compounds of Thai Honeys from 3 Different Honeybee Species.
Biomedical Activity and Related Volatile Compounds of Thai Honeys from 3 Different Honeybee Species.
Journal of food science (2015-09-01)
Praetinee Pattamayutanon, Sergio Angeli, Prodpran Thakeow, John Abraham, Terd Disayathanoowat, Panuwan Chantawannakul
ABSTRACT
This study investigated the effect of 3 factors (floral source, honeybee species, and postcollection processing) that influence the antibacterial activity, free radical reduction, and other biochemical compositions of different honey types typical of Thailand. Honey samples from 3 honeybee species (Apis mellifera, Apis cerana, and Apis dorsata) were obtained from 9 floral sources (longan, wild flower, lychee, coffee, sunflower, sesame, bitter bush, para-rubber, and manuka as a control) in different regions of Thailand. These samples were evaluated for both their total and nonperoxide antibacterial activity against 10 human pathogens by agar incorporation technique. Honey samples were further analyzed to evaluate the capacity for free radical-scavenging activity, total phenolic content, and the total flavonoid contents by the 2,2-diphenyl-1-picrylhydrazyl assay, Folin-Ciocalteu method, and aluminum chloride colorimetric assay, respectively. Furthermore, the volatile organic compounds (VOCs) of Thai honey samples were investigated by headspace solid-phase microextraction and gas chromatography-mass spectrometry analysis. Findings of this study suggest a strong correlation between floral origin and honeybee species on one hand, and differences in %Brix, total acidity, protein content, antimicrobial activities, free radical reduction, phenolic, and flavonoid contents on the other hand. Moreover, VOCs of wild and coffee honey types were remarkably different, depending on the floral source. Both honeys contained characteristics of VOCs, some of which are involved in antibacterial and antioxidant activities. Different origins and processing (floral source, honeybee species, and postcollection processing) of Thai honeys result in different antibacterial activities, physico-chemical properties, and aroma. Based on these findings, consumers of honey could select the type of honey based on their needs and preferred aroma.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Sigma-Aldrich
Methanol, anhydrous, 99.8%