Merck
  • Oligonucleotide biofunctionalization enhances endothelial progenitor cell adhesion on cobalt/chromium stents.
Oligonucleotide biofunctionalization enhances endothelial progenitor cell adhesion on cobalt/chromium stents.
Journal of biomedical materials research. Part A (2015-03-27)
Maria Chiara Barsotti, Tamer Al Kayal, Lorena Tedeschi, Dinuccio Dinucci, Paola Losi, Silverio Sbrana, Enrica Briganti, Rodorico Giorgi, Federica Chiellini, Rossella Di Stefano, Giorgio Soldani
ABSTRACT
As the endothelium still represents the ideal surface for cardiovascular devices, different endothelialization strategies have been attempted for biocompatibility and nonthrombogenicity enhancement. Since endothelial progenitor cells (EPCs) could accelerate endothelialization, preventing thrombosis and restenosis, the aim of this study was to use oligonucleotides (ONs) to biofunctionalize stents for EPC binding. In order to optimize the functionalization procedure before its application to cobalt-chromium (Co/Cr) stents, discs of the same material were preliminarily used. Surface aminosilanization was assessed by infrared spectroscopy and scanning electron microscopy. A fluorescent endothelial-specific ON was immobilized on aminosilanized surfaces and its presence was visualized by confocal microscopy. Fluorescent ON binding to porcine blood EPCs was assessed by flow cytometry. Viability assay was performed on EPCs cultured on unmodified, nontargeting ON or specific ON-coated discs; fluorescent staining of nuclei and F-actin was then performed on EPCs cultured on unmodified or specific ON-coated discs and stents. Disc biofunctionalization significantly increased EPC viability as compared to both unmodified and nontargeting ON-coated surfaces; cell adhesion was also significantly increased. Stents were successfully functionalized with the specific ON, and EPC binding was confirmed by confocal microscopy. In conclusion, stent biofunctionalization for EPC binding was successfully achieved in vitro, suggesting its use to obtain in vivo endothelialization, exploiting the natural regenerative potential of the human body.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%