Merck
  • Ultrasound intensification suppresses the need of methanol excess during the biodiesel production with Lipozyme TL-IM.
Ultrasound intensification suppresses the need of methanol excess during the biodiesel production with Lipozyme TL-IM.
Ultrasonics sonochemistry (2015-05-03)
Preeti B Subhedar, Claudia Botelho, Artur Ribeiro, Rita Castro, Maria Alcina Pereira, Parag R Gogate, Artur Cavaco-Paulo
ABSTRACT
The synthesis of biodiesel from sunflower oil and methanol based on transesterification using the immobilized lipase from Thermomyces lanuginosus (Lipozyme TL-IM) has been investigated under silent conditions and under an ultrasound field. Ultrasound assisted process led to reduced processing time and requirement of lower enzyme dosage. We found for the first time that oil to methanol ratio of 1:3 was favored for the ultrasound assisted enzymatic process which is lower than that observed for the case of conventional stirring based approach (ratio of 1.4). Our results indicate that intensification provided by ultrasound suppresses the need of the excess of the methanol reactant during the enzymatic biodiesel production. Ultrasound assisted enzymatic biodiesel production is therefore a faster and a cleaner processes.
Sigma-Aldrich
Methyl palmitate, ≥97%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methyl stearate, ≥96%, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Sigma-Aldrich
Methyl stearate, ~99% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methyl oleate, technical grade, 70%
Sigma-Aldrich
Methyl oleate, 99%