Merck
  • An Organic Catalyst for Li-O2 Batteries: Dilithium Quinone-1,4-Dicarboxylate.
An Organic Catalyst for Li-O2 Batteries: Dilithium Quinone-1,4-Dicarboxylate.
ChemSusChem (2015-06-16)
Jia Liu, Stéven Renault, Daniel Brandell, Torbjörn Gustafsson, Kristina Edström, Jiefang Zhu
ABSTRACT
Solid organic electrocatalysts have hardly been tested in Li-O2 batteries. Here, a new solid organic electrocatalyst, dilithium quinone-1,4-dicarboxylate (Li2 C8 H2 O6 ) is presented, which is expected to overcome the shortcomings of inorganic catalysts. The function-oriented synthesis is low cost and low polluting. The electrocatalytic performance is evaluated by following the degradation of Li2 O2 during the charge process in a Li-O2 cell through in situ XRD and operando synchrotron radiation powder XRD (SR-PXD) measurements. The results indicate that the electrocatalytic activity of Li2 C8 H2 O6 is similar to that of commercial Pt. The Li2 O2 decomposition in a cell with Li2 C8 H2 O6 catalyst follows a pseudo-zero-order reaction, virtually without any side reactions. These results provide an insight into the development of new organic catalysts for the oxygen evolution reaction (OER) in Li-O2 batteries.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Lithium-7Li2 carbonate, ≥99 atom % 7Li, 99% (CP)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2,5-Dihydroxyterephthalic acid, 98%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Lithium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis