Merck
  • Telomerase regulates MYC-driven oncogenesis independent of its reverse transcriptase activity.
Telomerase regulates MYC-driven oncogenesis independent of its reverse transcriptase activity.
The Journal of clinical investigation (2015-04-22)
Cheryl M Koh, Ekta Khattar, Shi Chi Leow, Chia Yi Liu, Julius Muller, Wei Xia Ang, Yinghui Li, Guido Franzoso, Shang Li, Ernesto Guccione, Vinay Tergaonkar
ABSTRACT
Constitutively active MYC and reactivated telomerase often coexist in cancers. While reactivation of telomerase is thought to be essential for replicative immortality, MYC, in conjunction with cofactors, confers several growth advantages to cancer cells. It is known that the reactivation of TERT, the catalytic subunit of telomerase, is limiting for reconstituting telomerase activity in tumors. However, while reactivation of TERT has been functionally linked to the acquisition of several "hallmarks of cancer" in tumors, the molecular mechanisms by which this occurs and whether these mechanisms are distinct from the role of telomerase on telomeres is not clear. Here, we demonstrated that first-generation TERT-null mice, unlike Terc-null mice, show delayed onset of MYC-induced lymphomagenesis. We further determined that TERT is a regulator of MYC stability in cancer. TERT stabilized MYC levels on chromatin, contributing to either activation or repression of its target genes. TERT regulated MYC ubiquitination and proteasomal degradation, and this effect of TERT was independent of its reverse transcriptase activity and role in telomere elongation. Based on these data, we conclude that reactivation of TERT, a direct transcriptional MYC target in tumors, provides a feed-forward mechanism to potentiate MYC-dependent oncogenesis.
Sigma-Aldrich
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Estradiol, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-TRF2 Antibody, clone 4A794, clone 4A794, Upstate®, from mouse
Pricing and availability is not currently available.