Merck
  • Selective determination of COXIBs in environmental water samples by mixed-mode solid phase extraction and liquid chromatography quadrupole time-of-flight mass spectrometry.
Selective determination of COXIBs in environmental water samples by mixed-mode solid phase extraction and liquid chromatography quadrupole time-of-flight mass spectrometry.
Journal of chromatography. A (2015-10-18)
Sara Triñanes, Maria Carmen Casais, Maria Carmen Mejuto, Rafael Cela
ABSTRACT
The development and performance evaluation of a method for the simultaneous determination of six pharmaceuticals belonging to the class of non-steroidal anti-inflammatory drugs (NSAIDs) which present high selectivity for the cyclooxygenase (COX)-2 isoform of COX (COXIBs) in environmental waters are presented. The method involves an off-line mixed mode (reversed-phase and strong anionic exchange) solid phase extraction (SPE) for the selective concentration of COXIBs in combination with liquid chromatography (LC) quadrupole time-of-flight (QTOF) mass spectrometry (MS). The use of a strong anionic exchange sorbent (Oasis MAX) led to a significant reduction of matrix effects, during electrospray ionization (ESI), in comparison with results obtained for mixed mode weak anionic exchange sorbent (Oasis Wax) and polymeric reversed phase sorbents (Oasis HLB and Strata X). The developed method attained limits of quantification (LOQs) between 0.01 and 0.20 ng L(-1) for river water and effluent wastewater and, for influent wastewater this limits were ranged between 0.03 and 0.45 ng L(-1). Among the pharmaceuticals investigated, two of them (celecoxib and etoricoxib) were detected at low levels (ppt) in real samples of treated and raw wastewaters, and two metabolites were also found; the carboxylated celecoxib and the hydroxylated etoricoxib.
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Valdecoxib, ≥98% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%