Merck
  • Effective Inhibition of Bone Morphogenetic Protein Function by Highly Specific Llama-Derived Antibodies.
Effective Inhibition of Bone Morphogenetic Protein Function by Highly Specific Llama-Derived Antibodies.
Molecular cancer therapeutics (2015-09-10)
Silvia Calpe, Koen Wagner, Mohamed El Khattabi, Lucy Rutten, Cheryl Zimberlin, Edward Dolk, C Theo Verrips, Jan Paul Medema, Hergen Spits, Kausilia K Krishnadath
ABSTRACT
Bone morphogenetic proteins (BMP) have important but distinct roles in tissue homeostasis and disease, including carcinogenesis and tumor progression. A large number of BMP inhibitors are available to study BMP function; however, as most of these antagonists are promiscuous, evaluating specific effects of individual BMPs is not feasible. Because the oncogenic role of the different BMPs varies for each neoplasm, highly selective BMP inhibitors are required. Here, we describe the generation of three types of llama-derived heavy chain variable domains (VHH) that selectively bind to either BMP4, to BMP2 and 4, or to BMP2, 4, 5, and 6. These generated VHHs have high affinity to their targets and are able to inhibit BMP signaling. Epitope binning and docking modeling have shed light into the basis for their BMP specificity. As opposed to the wide structural reach of natural inhibitors, these small molecules target the grooves and pockets of BMPs involved in receptor binding. In organoid experiments, specific inhibition of BMP4 does not affect the activation of normal stem cells. Furthermore, in vitro inhibition of cancer-derived BMP4 noncanonical signals results in an increase of chemosensitivity in a colorectal cancer cell line. Therefore, because of their high specificity and low off-target effects, these VHHs could represent a therapeutic alternative for BMP4(+) malignancies.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Human BMP2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human BMP-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human BMP-4 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Bone Morphogenetic Protein 4 human, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Bone Morphogenetic Protein 4 human, BMP-4, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Human BMP-5 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
BMP-4 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
BMP-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture