Merck
  • A microfluidic immunoassay platform for the detection of free prostate specific antigen: a systematic and quantitative approach.
A microfluidic immunoassay platform for the detection of free prostate specific antigen: a systematic and quantitative approach.
The Analyst (2015-05-20)
Narayanan Madaboosi, Ruben R G Soares, Virginia Chu, João Pedro Conde
ABSTRACT
As a leading cause of cancer-related deaths in men globally, prostate cancer (PCa) demands immense attention for theranostic purposes. There is an increasing need for the development of rapid, sensitive, economical, miniaturized and multiplexable assays. Towards this goal, we present a systematic approach for the optimisation of a microfluidic sandwich immunoassay, which can be applied as a generic biosensor platform for PCa detection. Prostate specific antigen (PSA) was used as the model biomarker, and its free form was captured using commercially available antibodies and detected using chemiluminescence, both in spiked buffer and matrix solutions. Along with the optimisation of surface chemistry and microfluidic parameters, we report a bio-affinity amplification strategy based on biotin-streptavidin chemistry to bring the limits of detection for free-PSA from 21.4 ng mL(-1) down to 2.7 ng mL(-1), within the clinically relevant range. An estimate of the surface coverage and simulations of the interactions taking place in the microfluidic biosensor during the assay are also presented. This novel platform using a simple passive adsorption-based bio-affinity strategy, when coupled with multiplexing and integrated detection, can serve as a promising point-of-care diagnostic tool for PCa.
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Luminol, 97%