Merck
  • Efficacy of humidity retention bags for the reduced adsorption and improved cleaning of tissue proteins including prion-associated amyloid to surgical stainless steel surfaces.
Efficacy of humidity retention bags for the reduced adsorption and improved cleaning of tissue proteins including prion-associated amyloid to surgical stainless steel surfaces.
Biofouling (2015-08-13)
T J Secker, H E Pinchin, R C Hervé, C W Keevil
ABSTRACT
Increasing drying time adversely affects attachment of tissue proteins and prion-associated amyloid to surgical stainless steel, and reduces the efficacy of commercial cleaning chemistries. This study tested the efficacy of commercial humidity retention bags to reduce biofouling on surgical stainless steel and to improve subsequent cleaning. Surgical stainless steel surfaces were contaminated with ME7-infected brain homogenates and left to dry for 15 to 1,440 min either in air, in dry polythene bags or within humidity retention bags. Residual contamination pre/post cleaning was analysed using Thioflavin T/SYPRO Ruby dual staining and microscope analysis. An increase in biofouling was observed with increased drying time in air or in sealed dry bags. Humidity retention bags kept both protein and prion-associated amyloid minimal across the drying times both pre- and post-cleaning. Therefore, humidity bags demonstrate a cheap, easy to implement solution to improve surgical instrument reprocessing and to potentially reduce associated hospital acquired infections.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)