Merck
  • Dried distillers grains with solubles do not always cause late blowing in baby Swiss cheese.
Dried distillers grains with solubles do not always cause late blowing in baby Swiss cheese.
Journal of dairy science (2015-10-12)
V Manimanna Sankarlal, E D Testroet, D C Beitz, S Clark
ABSTRACT
Late blowing in Swiss cheese, a result of unwanted gas production, is unacceptable to consumers and causes economic loss to manufacturers. Cheese processors have raised concerns that feeding dried distillers grains with solubles (DDGS) to cows leads to this defect, in part because of clostridial spores. We hypothesized that spores in DDGS would affect the quality of milk and baby Swiss cheese by promoting late-blowing defects. Thirty healthy multiparous and mid-lactation Holstein cows were fed total mixed ration containing DDGS (0, 10, and 20%; 10 cows per treatment group) by dietary dry matter in a 3×3 Latin square design. One complete milking from all cows within a treatment was collected and pooled for baby Swiss cheese, twice within each month of the 3-mo study. Additionally, individual milk samples from the 3 milkings of one day were collected weekly for proximate analysis. Incubation in reinforced clostridial medium-lactate medium tubes inoculated with milk, cheese, total mixed ration, or manure showed gas formation. Conversely, the DDGS used in our study did not contain gas-producing, spore-forming bacteria. Feeding 20% DDGS decreased milk fat percent and increased the solids nonfat, protein, and lactose percent of milk. After 60 d of ripening, baby Swiss cheese had typical propionic acid Swiss cheese aroma. Regardless of dietary treatment, pinholes, slits, splits, cracks, or a combination of these, were seen throughout most cheeses. Feeding of DDGS increased the amount of long-chain unsaturated fatty acids and decreased short-chain and most medium-chain fatty acids in the baby Swiss cheese. Although feeding cows diets with DDGS modified milk composition, and subsequently cheese composition, DDGS was not a source for gas-producing, spore-forming bacteria or for quality defects in Swiss cheese. Rather, the gas-producing, spore-forming bacteria likely originated from the environment or the cows themselves.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Cysteine, Vetec, reagent grade, 97%
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)