Merck
Combined IL-15 and IL-12 drives the generation of CD34
Oncoimmunology (2015-07-04)
Jeannette Cany, Anniek B van der Waart, Jan Spanholtz, Marleen Tordoir, Joop H Jansen, Robbert van der Voort, Nicolaas M Schaap, Harry Dolstra
ABSTRACT
Adoptive transfer of allogeneic natural killer (NK) cells represents a promising treatment approach against cancer, including acute myeloid leukemia (AML). Previously, we reported a cytokine-based culture method for the generation of NK cell products with high cell number and purity. In this system, CD34
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
IL-15 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-15 human, >97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture