Merck
  • Urinary extracellular vesicles as reservoirs of altered proteins during the pathogenesis of polycystic kidney disease.
Urinary extracellular vesicles as reservoirs of altered proteins during the pathogenesis of polycystic kidney disease.
Proteomics. Clinical applications (2015-03-11)
Gabriella Pocsfalvi, Delfin A A Raj, Immacolata Fiume, Annalisa Vilasi, Francesco Trepiccione, Giovambattista Capasso
ABSTRACT
Recent findings indicate that urinary extracellular vesicles (EVs) might reflect the pathophysiological state of urinary system; and that EVs-induced ciliary signaling is a possible mechanism of intercellular communication within the tract. Here, we aimed to analyze the protein expression of urinary EVs during autosomal dominant polycystic kidney disease (ADPKD). EVs were isolated from pooled urine samples of healthy control and ADPKD patients at two different stages of the disease and under tolvaptan treatment using the double-cushion ultracentrifugation method. Proteins were identified and quantified by iTRAQ and multidimensional protein identification technology (MudPIT)-based quantitative proteomics. Quantitative analyses identified 83 differentially expressed EV proteins. Many of these have apical membrane origin and are involved in signal transduction pathways of primary cilia, Ca(2+) -activated signaling, cell-cycle regulation, and cell differentiation. The reduced AQP-2 and the increased APO-A1 levels observed in all ADPKD-affected groups may reflects the impaired renal concentrating capability of these patients and correlated with estimated glomerular filtration rate decline. The levels of some upregulated proteins involved in Ca(2+) -activated signaling declined upon tolvaptan treatment. The results obtained suggest that the quantitative proteomics of urinary EVs might be useful to monitor proteins difficult to access noninvasively, and thus advance our understanding of urinary tract physiology and pathology.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
SAFC
BIS-TRIS
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
BIS-TRIS
SAFC
MOPS
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Nitrogen, ≥99.998%