Merck
  • New approach for the clinical monitoring of 25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 by ultra high performance liquid chromatography with MS/MS based on the standard reference material 972.
New approach for the clinical monitoring of 25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 by ultra high performance liquid chromatography with MS/MS based on the standard reference material 972.
Journal of separation science (2013-10-15)
Jiří Plíšek, Lenka Kujovská Krčmová, Jana Aufartová, Tanausú V Morales, Sarah M Esponda, Roman Oros, Eva Kasalová, Jose J Santana-Rodriguez, Luboš Sobotka, Petr Solich, Dagmar Solichová
ABSTRACT
Biomarkers, 25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 , are important indicators of the vitamin D general status and are monitored in several pathophysiological disorders, such as osteoporosis, diabetes, heart disease, etc. A novel ultra-HPLC with MS/MS methodology for the analysis of 25-hydroxyvitamin D derivatives coupled with a very simple and highly rapid sample preparation step was developed. Analytical parameters obtained showed linearity (R(2) ) above 0.999 for both vitamins with accuracies between 95.8 and 102%. The LODs were as low as 0.22 and 0.67 nmol/L for 25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 , respectively. Intra-assay precision (%RSD) was lower than 4.5%, and inter-assay precision (%RSD) was lower than 6.5%. The feasibility of the developed methodology to be applied in clinical routine analysis has been proved by its application in blood samples from non-agenarian patients, patients with familial hypercholesterolemia and patients suffering from age-related macular degeneration.
Sigma-Aldrich
Zinc sulfate monohydrate, purum p.a., ≥99.0% (KT)
Sigma-Aldrich
Zinc sulfate monohydrate, ≥99.9% trace metals basis