Merck
  • Screening UBQLN-2 in French frontotemporal lobar degeneration and frontotemporal lobar degeneration-amyotrophic lateral sclerosis patients.
Screening UBQLN-2 in French frontotemporal lobar degeneration and frontotemporal lobar degeneration-amyotrophic lateral sclerosis patients.
Neurobiology of aging (2013-04-16)
Serena Lattante, Isabelle Le Ber, Agnès Camuzat, Jérémie Pariente, Alexis Brice, Edor Kabashi
ABSTRACT
The ubiquilin-2 gene (UBQLN-2) is the only amyotrophic lateral sclerosis (ALS)-related gene mapping on the X chromosome. Mutations in the PXX domain of UBQLN-2 have been first described in ALS patients with a mutational frequency of 2.6% in familial ALS cases with no evidence of male-to-male transmission. Different populations have been further tested with mutations largely distributed in the gene and lower frequency of positive cases. To determine the genetic contribution of UBQLN-2 in frontotemporal lobar degeneration (FTLD) and FTLD-ALS, we screened a cohort of 136 French patients, identifying a missense variant (c.1006A>G; p.T336A) in 1 FTLD patient whose biological relevance to disease is questionable. We conclude that UBQLN-2 mutations related to ALS/FTLD are extremely rare in French FTLD and FTLD-ALS patients and should not be analyzed systematically.