Merck
  • Mutations in CNNM4 cause recessive cone-rod dystrophy with amelogenesis imperfecta.
Mutations in CNNM4 cause recessive cone-rod dystrophy with amelogenesis imperfecta.
American journal of human genetics (2009-02-10)
Bozena Polok, Pascal Escher, Aude Ambresin, Eliane Chouery, Sylvain Bolay, Isabelle Meunier, Francis Nan, Christian Hamel, Francis L Munier, Bernard Thilo, André Mégarbané, Daniel F Schorderet
ABSTRACT
Cone-rod dystrophies are inherited dystrophies of the retina characterized by the accumulation of deposits mainly localized to the cone-rich macular region of the eye. Dystrophy can be limited to the retina or be part of a syndrome. Unlike nonsyndromic cone-rod dystrophies, syndromic cone-rod dystrophies are genetically heterogeneous with mutations in genes encoding structural, cell-adhesion, and transporter proteins. Using a genome-wide single-nucleotide polymorphism (SNP) haplotype analysis to fine map the locus and a gene-candidate approach, we identified homozygous mutations in the ancient conserved domain protein 4 gene (CNNM4) that either generate a truncated protein or occur in highly conserved regions of the protein. Given that CNNM4 is implicated in metal ion transport, cone-rod dystrophy and amelogenesis imperfecta may originate from abnormal ion homeostasis.